Learn More
Medchemexpress LLC Sodium formate-13C,d1 | 1215684-17-5 | ≥98.0% | 71.01 g·mol⁻¹ | 13CHDNaO2 | 10 MG
SDP
Catalog No. 5000208282
Encompass_Preferred
Sodium formate-13C,d1 is a stable-isotope labeled sodium formate used as an internal standard and tracer in analytical applications such as mass spectrometry and NMR. It is supplied as a white to off-white solid with high chemical purity and is provided in small quantities for analytical work.
  • Stable-isotope labeled (13C and d1) sodium formate for tracer applications.
  • High chemical purity suitable for mass spectrometry and NMR.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Includes SDS and certificate of analysis for quality and safety verification.
Catalog No. 50-002-08282 Supplier Medchemexpress LLC Supplier No. HYW088065S110MG
Sodium formate-13C,d1 is a stable-isotope labeled sodium formate used as an internal standard and tracer in analytical applications such as mass spectrometry and NMR. It is supplied as a white to off-white solid with high chemical purity and is provided in small quantities for analytical work.
  • Stable-isotope labeled (13C and d1) sodium formate for tracer applications.
  • High chemical purity suitable for mass spectrometry and NMR.
  • Supplied as a white to off-white solid for easy handling and storage.
  • Includes SDS and certificate of analysis for quality and safety verification.