Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium formate-13C,d1 | 1215684-17-5 | 98.0% | 71.01 g/mol | 13CHDNaO2 | 25 MG
SDP

Catalog No. 5000208641
Encompass_Preferred

Sodium formate-13C,d1 is a stable-isotope labeled form of sodium formate in which the formyl carbon is 13C-labeled and the hydrogen is deuterated (d1). It is supplied as a solid reagent for use as an isotopic tracer and as a reducing reagent in biochemical and catalytic studies, and for material synthesis.

  • Stable-isotope labeled: 13C and d1
  • Purity: 98.0%
  • Molecular weight: 71.01 g/mol
  • Chemical formula: 13CHDNaO2
  • CAS number: 1215684-17-5
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg

Catalog No. 50-002-08641 Supplier Medchemexpress LLC Supplier No. 25MGHYW088065S1
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium formate-13C,d1 is a stable-isotope labeled form of sodium formate in which the formyl carbon is 13C-labeled and the hydrogen is deuterated (d1). It is supplied as a solid reagent for use as an isotopic tracer and as a reducing reagent in biochemical and catalytic studies, and for material synthesis.

  • Stable-isotope labeled: 13C and d1
  • Purity: 98.0%
  • Molecular weight: 71.01 g/mol
  • Chemical formula: 13CHDNaO2
  • CAS number: 1215684-17-5
  • Available pack sizes: 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.