Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium formate | 141-53-7 | MFCD00013101 | 99.0% | 68.01 g/mol | CHNaO2 | 100 G
SDP

Catalog No. 5000208357
Encompass_Preferred

Sodium formate is an inorganic salt used as a biochemical assay reagent and as a reducing agent in catalytic and sample-preparation applications. It promotes deprotonation of ligands at elevated temperatures and supplies reducing equivalents in catalytic oxidations.

  • Purity 99.0%.
  • Chemical formula CHNaO2; molecular weight 68.01 g/mol.
  • CAS number 141-53-7.
  • Available in 100 g pack size for laboratory use.
  • Common application as a reducing agent and reagent in biochemical assays.

Catalog No. 50-002-08357 Supplier Medchemexpress LLC Supplier No. HYW088065100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium formate is an inorganic salt used as a biochemical assay reagent and as a reducing agent in catalytic and sample-preparation applications. It promotes deprotonation of ligands at elevated temperatures and supplies reducing equivalents in catalytic oxidations.

  • Purity 99.0%.
  • Chemical formula CHNaO2; molecular weight 68.01 g/mol.
  • CAS number 141-53-7.
  • Available in 100 g pack size for laboratory use.
  • Common application as a reducing agent and reagent in biochemical assays.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.