Learn More
Medchemexpress LLC Sodium formate | 141-53-7 | MFCD00013101 | 99.0% | 68.01 g/mol | CHNaO2 | 25 G
SDP
Catalog No. 5000209198
Encompass_Preferred
Sodium formate is a white, crystalline biochemical assay reagent used as a deprotonating and mild reducing agent in catalytic reactions and analytical assays. It is commonly employed to form zinc oxide layers on substrates and to control oxidation processes in methane-to-methanol catalysis.
  • High purity 99.0% suitable for analytical and assay use.
  • Cas number 141-53-7 and molecular weight 68.01 g/mol.
  • Solid powder, commonly supplied as 25 G packages.
  • Functions as a deprotonating agent and mild reducing agent.
  • Useful in materials preparation for forming zinc oxide layers and in catalytic oxidation control.
Catalog No. 50-002-09198 Supplier Medchemexpress LLC Supplier No. 25GHYW088065
Sodium formate is a white, crystalline biochemical assay reagent used as a deprotonating and mild reducing agent in catalytic reactions and analytical assays. It is commonly employed to form zinc oxide layers on substrates and to control oxidation processes in methane-to-methanol catalysis.
  • High purity 99.0% suitable for analytical and assay use.
  • Cas number 141-53-7 and molecular weight 68.01 g/mol.
  • Solid powder, commonly supplied as 25 G packages.
  • Functions as a deprotonating agent and mild reducing agent.
  • Useful in materials preparation for forming zinc oxide layers and in catalytic oxidation control.