Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium formate | 141-53-7 | MFCD00013101 | 99.0% | 68.01 g/mol | CHNaO2 | 250 G
SDP

Catalog No. 5000209441
Encompass_Preferred

Sodium formate is the sodium salt of formic acid supplied as a solid biochemical reagent and reducing agent. It has a listed purity of 99.0% and is commonly used in catalytic oxidations and biochemical assays; storage recommendations include sealed storage at 4°C, away from moisture.

  • High purity reagent: 99.0% purity suitable for analytical and assay use.
  • Solid form: supplied as a dry powder for easy handling and storage.
  • Reduces oxidation: provides reducing force in catalytic oxidation reactions.
  • Surface chemistry: can react with zinc salts to form zinc oxide layers on supports.
  • Multiple pack sizes: available in laboratory pack sizes, including 250 g.
  • Stable storage: store sealed at 4°C and protect from moisture; in solvent, -80°C (6 months) or -20°C (1 month) for short-term.

Catalog No. 50-002-09441 Supplier Medchemexpress LLC Supplier No. 250GHYW088065
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium formate is the sodium salt of formic acid supplied as a solid biochemical reagent and reducing agent. It has a listed purity of 99.0% and is commonly used in catalytic oxidations and biochemical assays; storage recommendations include sealed storage at 4°C, away from moisture.

  • High purity reagent: 99.0% purity suitable for analytical and assay use.
  • Solid form: supplied as a dry powder for easy handling and storage.
  • Reduces oxidation: provides reducing force in catalytic oxidation reactions.
  • Surface chemistry: can react with zinc salts to form zinc oxide layers on supports.
  • Multiple pack sizes: available in laboratory pack sizes, including 250 g.
  • Stable storage: store sealed at 4°C and protect from moisture; in solvent, -80°C (6 months) or -20°C (1 month) for short-term.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.