Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium formate, 99%

Catalog No. AC148230010
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC148230010 1 kg Plastic bottle
AC148230051 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC148230010 Supplier Thermo Scientific Chemicals Supplier No. 148230010
  • $71.50
    $60.65 / Each of 1

    Save $10.85

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 141-53-7 | CHNaO2 | 68.007 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141-53-7
Molecular Formula CHNaO2
Molecular Weight (g/mol) 68.007
MDL Number MFCD00013101
InChI Key HLBBKKJFGFRGMU-UHFFFAOYSA-M
Synonym sodium formate, formic acid, sodium salt, salachlor, formic acid sodium salt, formic acid, na salt, sodium formate, refined, sodium formiate, mravencan sodny czech, ccris 1037, hsdb 744
PubChem CID 2723810
ChEBI CHEBI:62965
IUPAC Name sodium;formate
SMILES C(=O)[O-].[Na+]

Specifications

Melting Point 258.0°C to 261.0°C
Color White
Boiling Point 360.0°C
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula HCO2Na
Quantity 1 kg
Beilstein 02, 8
Fieser 8458
Merck Index 15, 8756
Solubility Information Solubility in water: 550g/L (20°C). Other solubilities: soluble in glycerol, slightly soluble in alcohol, insoluble in ether
Formula Weight 68.01
Percent Purity 0.99
Physical Form Crystalline Powder
Chemical Name or Material Sodium formate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.