Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium formate, Analytical Standard, 99.0-101.0 wt. % (NT), MilliporeSigma™ Supelco™

Catalog No. 111013075 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-3075 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-101-3075 Supplier MilliporeSigma Supelco Supplier No. 672531G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

CAS 141-53-7
Physical Form Neat
Linear Formula HCOONa
MDL Number MFCD00013101
Melting Point 259°C to 262°C
Synonym E237; Formic acid sodium salt
Solubility Soluble in water
Quantity 1 g
Formula Weight 68.01
Percent Purity 99.0 to 101.0 wt. % (NT)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.