Learn More
Medchemexpress LLC Sodium formate-d | 3996-15-4 | 99.8% | 69.01 | CDNaO2 | 1 G
SDP
Catalog No. 5000208284
Encompass_Preferred
Deuterium-labeled sodium formate used as a stable-isotope tracer and internal standard for analytical methods, and as a biochemical reagent in catalytic reactions. Supplied as a high-purity solid suitable for NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled sodium formate for use as tracer and internal standard.
  • Suitable for NMR, GC-MS, and LC-MS quantitation.
  • High purity: 99.8%.
  • Chemical formula CDNaO2; molecular weight 69.01.
  • Available as 1 g solid; soluble in water and DMSO.
  • Store sealed at 4°C away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-08284 Supplier Medchemexpress LLC Supplier No. HYW088065S21G
Deuterium-labeled sodium formate used as a stable-isotope tracer and internal standard for analytical methods, and as a biochemical reagent in catalytic reactions. Supplied as a high-purity solid suitable for NMR, GC-MS, and LC-MS applications.
  • Deuterium-labeled sodium formate for use as tracer and internal standard.
  • Suitable for NMR, GC-MS, and LC-MS quantitation.
  • High purity: 99.8%.
  • Chemical formula CDNaO2; molecular weight 69.01.
  • Available as 1 g solid; soluble in water and DMSO.
  • Store sealed at 4°C away from moisture; in solution store at -80°C (6 months) or -20°C (1 month).