Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium formate-d | 3996-15-4 | 99.8% | 69.01 | CDNaO2 | 500 MG
SDP

Catalog No. 5000208285
Encompass_Preferred

Sodium formate-d is the deuterium-labeled analog of sodium formate supplied as a solid stable-isotope reagent used as a tracer and internal standard in analytical chemistry and as a biochemical reagent in synthesis and catalytic reactions. It is provided with high purity and characterized solubility and storage recommendations.

  • Deuterium-labeled tracer for analytical quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High purity: 99.8%.
  • Molecular weight 69.01; chemical formula CDNaO2.
  • Solubility: water ≥ 100 mg/mL; DMSO limited, may require warming.
  • Storage: store sealed at 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).
  • Supplied as a 500 mg solid.

Catalog No. 50-002-08285 Supplier Medchemexpress LLC Supplier No. HYW088065S2500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium formate-d is the deuterium-labeled analog of sodium formate supplied as a solid stable-isotope reagent used as a tracer and internal standard in analytical chemistry and as a biochemical reagent in synthesis and catalytic reactions. It is provided with high purity and characterized solubility and storage recommendations.

  • Deuterium-labeled tracer for analytical quantitation.
  • Suitable as an internal standard for NMR, GC-MS, and LC-MS.
  • High purity: 99.8%.
  • Molecular weight 69.01; chemical formula CDNaO2.
  • Solubility: water ≥ 100 mg/mL; DMSO limited, may require warming.
  • Storage: store sealed at 4°C; in solution keep at -80°C (6 months) or -20°C (1 month).
  • Supplied as a 500 mg solid.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.