Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium formate-d | 3996-15-4 | 99.8% | 69.01 g/mol | CDNaO2 | 100 MG
SDP

Catalog No. 5000208283
Encompass_Preferred

Deuterium-labeled sodium formate supplied as a stable-isotope reagent for analytical and biochemical applications. High isotopic and chemical purity makes it suitable as an internal standard for mass spectrometry, an NMR reference, and for isotope-labeling studies.

  • Deuterium-labeled sodium formate (CDNaO2)
  • Purity 99.8%
  • CAS number 3996-15-4
  • Molecular weight 69.01 g/mol
  • Soluble in water and DMSO
  • Intended for use as internal standard, NMR reference, or isotopic tracer

Catalog No. 50-002-08283 Supplier Medchemexpress LLC Supplier No. HYW088065S2100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Deuterium-labeled sodium formate supplied as a stable-isotope reagent for analytical and biochemical applications. High isotopic and chemical purity makes it suitable as an internal standard for mass spectrometry, an NMR reference, and for isotope-labeling studies.

  • Deuterium-labeled sodium formate (CDNaO2)
  • Purity 99.8%
  • CAS number 3996-15-4
  • Molecular weight 69.01 g/mol
  • Soluble in water and DMSO
  • Intended for use as internal standard, NMR reference, or isotopic tracer

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.