Learn More
Medchemexpress LLC Sodium formononetin-3'-sulfonate | 949021-68-5 | 99.8% | 10 MM * 1 ML
SDP
Catalog No. 5000341079
Encompass_Preferred
Sodium formononetin-3'-sulfonate (Sul-F) is a water-soluble derivative of formononetin. Research indicates that Sul-F significantly prevents the elevation of ST-segment levels and decreases several serum markers associated with myocardial injury. It is also noted for its favorable water solubility, lipid-lowering, and liver-protection activities, making it suitable for various research applications.
  • Prevents elevation of ST-segment level.
  • Decreases creatine kinase-MB, lactate dehydrogenase, alanine aminotransferase, and cardiac troponin T levels.
  • Reduces myocardium necrosis scores.
  • Elevates expression of endogenous antioxidant defense enzymes.
  • Degrades lipid peroxidation products.
  • Improves energy metabolism of cardiac mitochondria.
  • Attenuates cardiocyte apoptosis.
  • Exhibits good lipid-lowering and liver-protection activities.
Catalog No. 50-003-41079 Supplier Medchemexpress LLC Supplier No. HY1306310MM/1ML
Sodium formononetin-3'-sulfonate (Sul-F) is a water-soluble derivative of formononetin. Research indicates that Sul-F significantly prevents the elevation of ST-segment levels and decreases several serum markers associated with myocardial injury. It is also noted for its favorable water solubility, lipid-lowering, and liver-protection activities, making it suitable for various research applications.
  • Prevents elevation of ST-segment level.
  • Decreases creatine kinase-MB, lactate dehydrogenase, alanine aminotransferase, and cardiac troponin T levels.
  • Reduces myocardium necrosis scores.
  • Elevates expression of endogenous antioxidant defense enzymes.
  • Degrades lipid peroxidation products.
  • Improves energy metabolism of cardiac mitochondria.
  • Attenuates cardiocyte apoptosis.
  • Exhibits good lipid-lowering and liver-protection activities.