Learn More
Medchemexpress LLC Sodium formononetin-3'-sulfonate | 949021-68-5 | 99.8% | C16H11NaO7S | 10MG
SDP
Catalog No. 5000259784
Encompass_Preferred
Sodium formononetin-3'-sulfonate is a water-soluble sulfonate derivative of formononetin intended for research applications. It is provided as a high-purity solid optimized for biochemical and cellular experiments where improved aqueous solubility and stability relative to the parent isoflavone are required. Storage and solubility guidance should be followed for best results.
  • High reported purity (≈99.8%).
  • Solid form with options for solution preparations.
  • Readily soluble in DMSO (100 mg/mL) and water (25 mg/mL) with ultrasonic aid.
  • Recommended storage: refrigerated sealed for solid; frozen in solvent for long-term storage.
  • Suitable for biochemical and cellular oxidative stress studies.
Catalog No. 50-002-59784 Supplier Medchemexpress LLC Supplier No. HY1306310MG
Sodium formononetin-3'-sulfonate is a water-soluble sulfonate derivative of formononetin intended for research applications. It is provided as a high-purity solid optimized for biochemical and cellular experiments where improved aqueous solubility and stability relative to the parent isoflavone are required. Storage and solubility guidance should be followed for best results.
  • High reported purity (≈99.8%).
  • Solid form with options for solution preparations.
  • Readily soluble in DMSO (100 mg/mL) and water (25 mg/mL) with ultrasonic aid.
  • Recommended storage: refrigerated sealed for solid; frozen in solvent for long-term storage.
  • Suitable for biochemical and cellular oxidative stress studies.