Learn More
Description
- Sodium Free Mineral Mixture
- For rodent diet
- Powder
Specifications
Specifications
| Product Type | Animal Diet |
| For Use With (Application) | Rodent diet |
| Quantity | 2 kg |
| Storage Requirements | Room Temperature |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.