Learn More
Selleck Chemical LLC Sodium Glucoheptonate Dihydrate S2587-25mg
SDP
Catalog No. 503110981
Encompass_Preferred
Sodium glucoheptonate is used for cleaning metal mercerizing paint stripping and aluminum etching
Catalog No. 50-311-0981 Supplier Selleck Chemical LLC Supplier No. S258725MG
Sodium glucoheptonate is used for cleaning metal mercerizing paint stripping and aluminum etching