Learn More
Apexbio Technology LLC SODIUM-GLUCONATE-100MG
SDP
Catalog No. 5000568075
Encompass_Preferred
Sodium Gluconate (CAS No 527-07-1) is widely studied in biomedical research due to its chelating properties and potential therapeutic applications Originating from gluconic acid it functions by impacting metal ion pathways crucial to cellular processes It exhibits in vitro activity in the micromolar range highlighting its efficacy in binding heavy metals such as calcium and iron This compound is commonly used in various applications including cell models and animal studies where it aids in drug screening efforts by modulating metal ion concentrations Its ability to stabilize ionic environments makes it valuable in experiments focused on cellular growth and metabolism Experimental concentrations typically depend on the specific research design enabling adaptability across different study parameters This flexibility ensures its continued relevance in probing the complex interactions of metal ions in biological systems
Catalog No. 50-005-68075 Supplier Apexbio Technology LLC Supplier No. B1836100
Sodium Gluconate (CAS No 527-07-1) is widely studied in biomedical research due to its chelating properties and potential therapeutic applications Originating from gluconic acid it functions by impacting metal ion pathways crucial to cellular processes It exhibits in vitro activity in the micromolar range highlighting its efficacy in binding heavy metals such as calcium and iron This compound is commonly used in various applications including cell models and animal studies where it aids in drug screening efforts by modulating metal ion concentrations Its ability to stabilize ionic environments makes it valuable in experiments focused on cellular growth and metabolism Experimental concentrations typically depend on the specific research design enabling adaptability across different study parameters This flexibility ensures its continued relevance in probing the complex interactions of metal ions in biological systems