Learn More
Sigma Aldrich Fine Chemicals Biosciences Sodium glycocholate hydrat25G
Catalog No. 502908033
Encompass
Sodium glycocholate hydrate is a conjugated bile salt and an ionic biologic detergent.
Catalog No. 50-290-8033 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. G713225G
Sodium glycocholate hydrate is a conjugated bile salt and an ionic biologic detergent.