Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Sodium glycolate | 2836-32-0 | MFCD00065358 | 98.0% | 98.03 g/mol | C2H3NaO3 | 10 G
SDP

Catalog No. 5000208538
Encompass_Preferred

Sodium glycolate is a high-purity inorganic salt used primarily as a metal chelating reagent in chemical and biochemical research. It is supplied as a solid and packaged for laboratory use with accompanying safety and quality documentation.

  • Purity: 98.0%.
  • Function: metal chelator for analytical and preparative applications.
  • Form: solid, water-soluble salt.
  • Molecular weight: 98.03 g/mol.
  • Identification: CAS 2836-32-0 and MFCD00065358.
  • Documentation: SDS and certificate of analysis typically provided.
  • Pack sizes: available in small laboratory quantities suitable for research.

Catalog No. 50-002-08538 Supplier Medchemexpress LLC Supplier No. HYW088011B10G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium glycolate is a high-purity inorganic salt used primarily as a metal chelating reagent in chemical and biochemical research. It is supplied as a solid and packaged for laboratory use with accompanying safety and quality documentation.

  • Purity: 98.0%.
  • Function: metal chelator for analytical and preparative applications.
  • Form: solid, water-soluble salt.
  • Molecular weight: 98.03 g/mol.
  • Identification: CAS 2836-32-0 and MFCD00065358.
  • Documentation: SDS and certificate of analysis typically provided.
  • Pack sizes: available in small laboratory quantities suitable for research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.