Learn More
Medchemexpress LLC Sodium heptane-1-sulfonate | 22767-50-6 | MFCD00007543 | 98.0% | 202.25 g·mol⁻¹ | C7H15NaO3S | 10 G
SDP
Catalog No. 5000302888
Encompass_Preferred
Sodium heptane-1-sulfonate is the sodium salt of heptane-1-sulfonic acid used as an ion-pairing reagent and buffer additive for biochemical research and reversed-phase HPLC separations. It is a water-soluble sulfonate that helps improve chromatographic retention and peak shape for ionic and polar analytes.
Catalog No. 50-003-02888 Supplier Medchemexpress LLC Supplier No. HYW01479210G
Sodium heptane-1-sulfonate is the sodium salt of heptane-1-sulfonic acid used as an ion-pairing reagent and buffer additive for biochemical research and reversed-phase HPLC separations. It is a water-soluble sulfonate that helps improve chromatographic retention and peak shape for ionic and polar analytes.