Learn More
Strem, An Ascensus Company CAS# 123334-23-6. 1g. Sodium hexachloroiridate(III) hydrate. MFCD00149194
SDP
Catalog No. 5090113163
Encompass
CAS# 123334-23-6. 1g. Sodium hexachloroiridate(III) hydrate. MFCD00149194. Molecular Weight: 473.91. Molecular Formula: Na3IrCl6∙XH2O. Color/form: greenish-brown solid Strem# 77-9000. http://www.strem.com/catalog/v/77-9000/
Catalog No. 50-901-13163 Supplier Strem, An Ascensus Company Supplier No. 7790001G
CAS# 123334-23-6. 1g. Sodium hexachloroiridate(III) hydrate. MFCD00149194. Molecular Weight: 473.91. Molecular Formula: Na3IrCl6∙XH2O. Color/form: greenish-brown solid Strem# 77-9000. http://www.strem.com/catalog/v/77-9000/