Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium hexafluoroacetylacetonate

Catalog No. 4361865G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
43-618-65G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 43-618-65G Supplier Sigma Aldrich Supplier No. 4361865G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Recently employed in the solid-phase synthesis of chromium α-diketonates, in the preparation of novel catalytic ruthenium diphosphine complexes and in the preparation of a series of new copper chemical vapor deposition (CVD) precursors.

Specifications

CAS 22466-49-5
Molecular Formula C5HF6NaO2
Linear Formula NaC5HF6O2
Quantity 5 g
MDL Number MFCD00156524
Synonym Sodium 1,1,1,5,5,5-hexafluoroacetylacetonate; Sodium hexafluoroacetylacetonate; Sodium hexafluoroacetylacetone
Molecular Weight (g/mol) 230.04
Percent Purity 97%
Biological Activity Respiratory system
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.