Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexafluoroaluminate, 97.8+%, Thermo Scientific Chemicals

Catalog No. p-7080447
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA8827536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA8827536 Supplier Thermo Scientific Chemicals Supplier No. 08827536
Only null left
Add to Cart

Sodium hexafluoroaluminate is a biochemical for proteomics research. It is used as a flux agent or bath material by aluminum smelters in the production of aluminum. It is also used in the production of synthetic resins for abrasives and in the manufacture of cutting or grinding discs. Minor uses are as a coloring agent (opacifier) in the glass, ceramic and pyrotechnic industries. It occurs naturally as the mineral cryolite, which is used extensively in the industrial production of aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13775-53-6
Molecular Formula Na3AlF6
MDL Number MFCD00003507
Synonym Cryolite; Aluminum sodium fluoride

Specifications

Melting Point 1,000°C
Physical Form Powder
Quantity 500 g
UN Number UN3260
Solubility Information Sparingly soluble in water(0.602g/L at 20°C).
Formula Weight 209.95
Percent Purity ≥97.8%
Odor Odorless
Density 2.90 g/mL
Chemical Name or Material Sodium hexafluoroaluminate
Hazard Category H332-H372
Hazard Statement GHS H Statement
H372-H302-H332
Causes damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P304+P340-P312-P314-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 237-410-6
RTECSNumber BD0075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.