Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hexafluoroaluminate, 99.4% (metals basis)

Catalog No. p-7050263
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3634530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA3634530 Supplier Thermo Scientific Chemicals Supplier No. 03634530

CAS: 13775-53-6

Mainly used as fluxing agent in the production of aluminum electrolysis, also used as additive to the wear-resisting of abrasive products, iron alloy and the flux of rimming steel, non-ferrous metal flux, casting deoxidizer, enamel of emulsifier, glass opal agent, welding flux and ceramics filler. Aluminum- metallurgy. This chemical is used as adhesives, cosolvent in enamel industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13775-53-6
MDL Number MFCD00003507
Synonym Aluminum sodium fluoride; Cryolite

Specifications

Melting Point 1,000°C
Physical Form Lump
Quantity 250 g
UN Number UN3260
Solubility Information Sparingly soluble in water 0.4g/L @ 20°C.
Formula Weight 209.95
Percent Purity 99.5%
Odor Odorless
Density 2.90 g/mL
Assay (metals basis)
Chemical Name or Material Sodium hexafluoroaluminate
Show More Show Less
Hazard Category H332-H372
Hazard Statement GHS H Statement
H372-H302-H332
Causes damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P260-P264b-P270-P271-P304+P340-P312-P314-P501c
DOTInformation Hazard Class: 9; Packaging Group: III
EINECSNumber 237-410-6
RTECSNumber BD0075000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.