Learn More
Matrix Scientific Sodium hexafluorophosphate, 21324-39-0, MFCD00011122, 25g
Supplier: Matrix Scientific 00653225G
Molecular Formula F6NaP, Purity 98%, Molecular Weight 167.95, Melting Point >200°
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.