Learn More
Gilson Company Inc Sodium Hexametaphosphate, 68915-31-1, MFCD00136045, 1 lb
Catalog No. NC0791039
Encompass_Preferred
Molecular Formula: 6 Na O18 P6, Molecular Weight: 611.766. Sodium Hexametaphosphate is a dry powder dispersion agent mixed into soil hydrometer solutions to prevent clay platelets from sticking together in the sedimentation solution. Required in hydrometer analysis of soils to determine silt and clay fractions.
Catalog No. NC0791039 Supplier Gilson Company Inc Supplier No. SA20
Molecular Formula: 6 Na O18 P6, Molecular Weight: 611.766. Sodium Hexametaphosphate is a dry powder dispersion agent mixed into soil hydrometer solutions to prevent clay platelets from sticking together in the sedimentation solution. Required in hydrometer analysis of soils to determine silt and clay fractions.