Learn More
Strem, An Ascensus Company CAS# 1070-89-9. 5g. Sodium hexamethyldisilazane, min. 95%. MFCD00009835
SDP
Catalog No. 5090109471
Encompass
CAS# 1070-89-9. 5g. Sodium hexamethyldisilazane, min. 95%. MFCD00009835. Molecular Weight: 183.38. Molecular Formula: [(CH3)3Si]2NNa. Color/form: off-white pwdr. Strem# 11-1500. http://www.strem.com/catalog/v/11-1500/
Catalog No. 50-901-09471 Supplier Strem, An Ascensus Company Supplier No. 1115005G
CAS# 1070-89-9. 5g. Sodium hexamethyldisilazane, min. 95%. MFCD00009835. Molecular Weight: 183.38. Molecular Formula: [(CH3)3Si]2NNa. Color/form: off-white pwdr. Strem# 11-1500. http://www.strem.com/catalog/v/11-1500/