Learn More
Medchemexpress LLC Sodium hexane-1-sulfonate | 2832-45-3 | MFCD00007542 | 98.0% | 188.22 g·mol⁻¹ | C6H13NaO3S | 25 G
SDP
Catalog No. 5000304196
Encompass_Preferred
Sodium hexane-1-sulfonate is an alkylsulfonate salt used as an ion-pairing reagent and buffer component in biochemical research and high-performance liquid chromatography (HPLC). It is supplied as a white to off-white solid, typically high purity, and should be stored sealed and protected from moisture.
  • Used as an ion-pairing reagent for HPLC and ion chromatography.
  • Functions as a buffer component in biochemical assays.
  • High purity suitable for analytical applications.
  • White to off-white solid for easy handling and weighing.
  • Stable when stored sealed, away from moisture.
Catalog No. 50-003-04196 Supplier Medchemexpress LLC Supplier No. 25GHYW016091
Sodium hexane-1-sulfonate is an alkylsulfonate salt used as an ion-pairing reagent and buffer component in biochemical research and high-performance liquid chromatography (HPLC). It is supplied as a white to off-white solid, typically high purity, and should be stored sealed and protected from moisture.
  • Used as an ion-pairing reagent for HPLC and ion chromatography.
  • Functions as a buffer component in biochemical assays.
  • High purity suitable for analytical applications.
  • White to off-white solid for easy handling and weighing.
  • Stable when stored sealed, away from moisture.