Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hexanitrocobaltate(III), Solstice

Catalog No. 60047376 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-376 25 g Glass bottle
60-047-375 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-047-376 Supplier Solstice Supplier No. 7160825G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

missing translation for 'validMainframeMsg'

Puriss. p.a., for the determination of K, ACS Reagent

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13600-98-1
Molecular Formula CoN6Na3O12
Molecular Weight (g/mol) 403.93
MDL Number MFCD00003512
InChI Key BZFWEBJDBATXME-UHFFFAOYSA-N
IUPAC Name trisodium hexanitrocobalttris(ylium)
SMILES [Na+].[Na+].[Na+].[O-][N+](=O)[Co+3]([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)([N+]([O-])=O)[N+]([O-])=O

Specifications

Melting Point 220°C
Quantity 25 g
Linear Formula Na3Co(NO2)6
UN Number UN1479
Formula Weight 403.94g/mol
Packaging Glass bottle
Chemical Name or Material Sodium hexanitrocobaltate(III)
Hazard Statement P220-P261-P280-P305 + P351 + P338-P342 + P311
DOTInformation 5.1 / PGII
RTECSNumber 237-077-7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.