Learn More
Medchemexpress LLC Sodium hippurate | 532-94-5 | MFCD00002693 | 99.9% | 201.16 g/mol | C9H8NNaO3 | 5 G
SDP
Catalog No. 5000302964
Encompass_Preferred
Sodium hippurate is the monosodium salt of hippuric acid supplied as a white to off-white solid reference material for biochemical and analytical research. It is used as a metabolite standard in chromatography and mass spectrometry workflows, and is provided at high purity suitable for laboratory assays.
  • High purity for analytical and research applications (≈99.9%).
  • Monosodium salt of hippuric acid used as a metabolite reference standard.
  • White to off-white solid, water soluble for convenient solution preparation.
  • Molecular formula C9H8NNaO3 and molecular weight 201.16 g/mol.
  • Available in small pack sizes suitable for lab use (5 g, 10 g, 50 g).
  • Suitable for use in chromatographic and mass spectrometric assays.
Catalog No. 50-003-02964 Supplier Medchemexpress LLC Supplier No. HYW0148415G
Sodium hippurate is the monosodium salt of hippuric acid supplied as a white to off-white solid reference material for biochemical and analytical research. It is used as a metabolite standard in chromatography and mass spectrometry workflows, and is provided at high purity suitable for laboratory assays.
  • High purity for analytical and research applications (≈99.9%).
  • Monosodium salt of hippuric acid used as a metabolite reference standard.
  • White to off-white solid, water soluble for convenient solution preparation.
  • Molecular formula C9H8NNaO3 and molecular weight 201.16 g/mol.
  • Available in small pack sizes suitable for lab use (5 g, 10 g, 50 g).
  • Suitable for use in chromatographic and mass spectrometric assays.