Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydrogen 4-Chlorophthalate (contains isomer and Phthalic Acid) 70.0+%, TCI America™
SDP

Catalog No. C026525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C026525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C026525G Supplier TCI America Supplier No. C026525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 56047-23-5
Molecular Formula C8H4ClNaO4
Molecular Weight (g/mol) 222.56
MDL Number MFCD00002495
InChI Key MWZMHSGEQHVWED-UHFFFAOYSA-M
Synonym sodium 4-chlorobenzene-1,2-dicarboxylic acid
PubChem CID 118797044
IUPAC Name sodium 2-carboxy-5-chlorobenzoate
SMILES [Na+].OC(=O)C1=CC=C(Cl)C=C1C([O-])=O

Specifications

Quantity 25 g
UN Number 2811
Formula Weight 222.56
Percent Purity ≥70.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Sodium Hydrogen 4-Chlorophthalate (contains isomer and Phthalic Acid)
EINECSNumber (3)-1412
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.