Learn More
Crescent Chemical Co Inc SODIUM HYDROGEN PHOSPHATE-2H2O
SDP
Catalog No. 50247912
Encompass_Preferred
Sodium hydrogen phosphate-2H2O analytical grade Buffering substance according to Sörensen. Biochemical and enzymatic standard, tested for use in tissue culture. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 30200.01 CAS 10028-24-7
Catalog No. 50-247-912 Supplier Crescent Chemical Co Inc Supplier No. SE30200.01
Sodium hydrogen phosphate-2H2O analytical grade Buffering substance according to Sörensen. Biochemical and enzymatic standard, tested for use in tissue culture. Size - 500G Storage Conditions - +15 °C TO +30 °C Catalog Number - 30200.01 CAS 10028-24-7