Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydrogen Selenite, purum p.a, ≥96.0% (RT), Solstice

Catalog No. 60047382 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-047-382 100 g Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-047-382 Supplier Solstice Supplier No. 71658100G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Purum p.a.,≥96.0% (RT)

  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-82-3
Molecular Formula Na2O3Se
Molecular Weight (g/mol) 172.95
MDL Number MFCD00003525
InChI Key BVTBRVFYZUCAKH-UHFFFAOYSA-L
Synonym sodium hydrogen selenite, sodium hydroselenite, sodium biselenite, selenious acid, monosodium salt, sodium selenite nahseo3, sodium hydrogenselenite, hydrogen sodium selenite, unii-him8pp9m79, sodium hydrogen trioxoselenite, hydrogen sodium selenium oxide
PubChem CID 23669629
IUPAC Name sodium;hydrogen selenite
SMILES [Na+].[Na+].[O-][Se]([O-])=O

Specifications

Quantity 100 g
Linear Formula NaHSeO3
UN Number UN2630
Formula Weight 150.96g/mol
Percent Purity ≥96.0% (RT)
Packaging Poly bottle
Chemical Name or Material Sodium hydrogen selenite
Hazard Statement P261-P273-P301 + P310-P311-P501
DOTInformation 6.1 / PGI
RTECSNumber 231-966-3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.