Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Sodium hydrogen sulfate monohydrate

Catalog No. 769339500G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-933-9500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 76-933-9500G Supplier Sigma Aldrich Supplier No. 769339500G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

The monohydrate of sodium hydrogen sulfate finds application as catalyst in organic reactions such as esterification of carboxylic acids and alcohols.

Specifications

CAS 10034-88-5
Quantity 500 g
Molecular Formula HNaO4S -+ H2O
Linear Formula NaHSO4 -+ H2O
MDL Number MFCD00149210
Synonym Sodium bisulfate monohydrate; Sodium hydrogen sulfate monohydrate
Molecular Weight (g/mol) 138.08
Percent Purity ≥99% (T)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.