Learn More
Cayman Chemical Sodium Hydrogen Sulfide hydr
SDP
Catalog No. 502888703
Encompass_Preferred
A H2S donor; commonly used in cellular and whole animal experimental systems; has been used to suggest that H2S promotes neutrophil migration, reduces airway inflammation, and protects neurites, heart, and intestine from chemical or ischemic-reperfusion damage
Catalog No. 50-288-8703 Supplier Cayman Chemical Supplier No. 100125551 G
A H2S donor; commonly used in cellular and whole animal experimental systems; has been used to suggest that H2S promotes neutrophil migration, reduces airway inflammation, and protects neurites, heart, and intestine from chemical or ischemic-reperfusion damage