Learn More
Aqua Solutions SODIUM HYDROX 4N TOC/IC 20L
SDP
Catalog No. NC0292915
Encompass_Preferred
Sodium Hydroxide 4.0 Normal Aqueous Solution N.i.s.t. Traceable (suitable For Toc Analysis)
Catalog No. NC0292915 Supplier Aqua Solutions Supplier No. 865720LNP
Sodium Hydroxide 4.0 Normal Aqueous Solution N.i.s.t. Traceable (suitable For Toc Analysis)