Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 0.01 N Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18613520 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
Packaging:
Cubitainer
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-613-520 20 L Cubitainer
18-613-517 500 mL Poly Bottle
18-613-518 1 L Poly Bottle
18-613-519 4 L Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-613-520 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S32020LT
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1310-73-2 | 40.00 g/mol

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5

Specifications

Concentration 0.01 N ± 0.0002 N
Quantity 20 L
Molecular Weight (g/mol) 40.00
Percent Purity 5% ±0.25%
Grade Chemical Solution
Packaging Cubitainer
Chemical Name or Material Sodium Hydroxide Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.