Learn More
Hach Company Sodium Hydroxide Standard Solution, 0.0227 N, 1 L
Catalog No. 501994194 Shop All Hach Company Products
Encompass
For carbon dioxide determination by buret titration. Hach method 8233. 1 liter bottle.
Catalog No. 50-199-4194 Supplier Hach Company Supplier No. 19253
For carbon dioxide determination by buret titration. Hach method 8233. 1 liter bottle.