Learn More
Hach Company Sodium Hydroxide Standard Solution, 0.020 N (N/50), 1 L
Supplier:  Hach Company 19353
Encompass
For acidity determination by buret titration. Hach methods 8219 (methyl orange) and 8010 (phenolphthalein). 1 liter bottle
Catalog No. 50-199-4195