Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Sodium Hydroxide 0.1 Normal NIST Traceable Solution (1L)
SDP

Catalog No. NC9820369
Encompass_Preferred

Sodium Hydroxide 0.1 Normal NIST Traceable Solution (1L)

Catalog No. NC9820369 Supplier Aqua Solutions Supplier No. 86251L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Hydroxide 0.1 Normal NIST Traceable Solution (1L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.