Learn More
Hach Company Sodium Hydroxide Digital Titrator Cartridge, 0.1600 N
Supplier:  Hach Company 1437701
Encompass
For acidity determination by Digital Titration, 10 - 160 mg/L CaCO3
Catalog No. 50-199-5008