Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium hydroxide, 0.1N, Macron Fine Chemicals™

Catalog No. MK61461 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
cubetainer
narrow mouth polyethylene bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MK-6146-1 1 L narrow mouth polyethylene bottle
MK-6146-4 4 L cubetainer
MK-6146-20 20 L cubetainer
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MK-6146-1 Supplier Avantor Supplier No. 614660
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color White
Density 2.13g/cm3
Quantity 1 L
Formula Weight 39.99
Packaging narrow mouth polyethylene bottle
Physical Form Liquid
Chemical Name or Material Sodium hydroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.