Learn More
Hach Company Sodium Hydroxide Digital Titrator Cartridge, 0.9274 N
Supplier:  Hach Company 1484201
Encompass
For volatile acids determination by Digital Titration, 100 - 2400 mg/L Acetic Acid
Catalog No. 50-199-5133