Learn More
Hach Company Sodium Hydroxide Standard Solution, 0.075 N, 100 mL MDB
Supplier:  Hach Company 2498032
Encompass
For total exchangeable acidity determination in soils. Used in test kit 2496000.
Catalog No. 50-199-6093