Learn More
Aqua Solutions SODIUM HYDROXIDE 1.0 NORM 4L
SDP
Catalog No. NC2213462
Encompass_Preferred
SODIUM HYDROXIDE 1.0 NORMAL NIST TRACEABLE(4LG) 4L
This item is not returnable. View return policy
Catalog No. NC2213462 Supplier Aqua Solutions Supplier No. 86504LG
SODIUM HYDROXIDE 1.0 NORMAL NIST TRACEABLE(4LG) 4L