Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 1.00 Normal, Reagents

Catalog No. 01336260
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
10 L
20 L
200 L
Packaging:
Cubitainer
Poly Bottle
Poly Drum
Catalog No. Quantity Packaging
01-336-260 500 mL Poly Bottle
01-336-266 10 L Cubitainer
01-336-265 1 L Poly Bottle
01-336-264 200 L Poly Drum
01-336-263 20 L Cubitainer
01-336-262 4 L Poly Bottle
01-336-261 4 L Cubitainer
7 options

Catalog No. 01-336-260

Supplier: Reagents Holdings Llc C7318500500A

Only null left
Add to Cart
Add to Cart

Sodium Hydroxide, 1.00 Normal, Reagent, for general laboratory use. Manufactured in ISO 9001 facility. Accredited to ISO 17025, NIST Traceable.

Chemical Identifiers

CAS 7732-18-5 , 1310-73-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Lye, Caustic soda
IUPAC Name water
SMILES O

Specifications

pH >13
Color Colorless
Boiling Point 100°C
Quantity 500 mL
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.044
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide, 1.00 Normal
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.