Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Sodium Hydroxide 1.0N NIST Traceable Solution (4L)
SDP

Catalog No. NC9893675
Encompass_Preferred

Sodium Hydroxide 1.0N NIST Traceable Solution (4L)

Catalog No. NC9893675 Supplier Aqua Solutions Supplier No. 86504LC
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sodium Hydroxide 1.0N NIST Traceable Solution (4L)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.