Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 1.0N Volumetric Solution, Macron Fine Chemicals™

Catalog No. MK46934 Shop All Avantor Products
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MK-4693-4 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MK-4693-4 Supplier Avantor Supplier No. 469301
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Certified 1.0N sodium hydroxide volumetric solution, traceable to NIST standards for precision laboratory and manufacturing applications.

Chemical Identifiers

CAS 1310-73-2
Molecular Formula HNaO
Molecular Weight (g/mol) 39.997
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
Synonym sodium hydroxide, caustic soda, sodium hydrate, white caustic, soda lye, aetznatron, ascarite, sodium hydroxide na oh, sodium hydroxide solution, soda, caustic
PubChem CID 14798
ChEBI CHEBI:32145
IUPAC Name sodium;hydroxide
SMILES [OH-].[Na+]

Specifications

Color Colorless
Quantity 4 L
Grade Volumetric
Normality 0.995 to 1.005 N
Packaging HDPE or Glass Bottle
Physical Form Liquid
Chemical Name or Material Sodium Hydroxide
storageNote1 Protect from air to avoid absorption of carbon dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.