Learn More
Aqua Solutions SODIUM HYDROXIDE 1.500N
SDP
Catalog No. NC9480421
Encompass_Preferred
Sodium Hydroxide 1.5 Normal solution;6%; Clear colorless solution;4L; CAS: 1310-73-2; Store in a cool dry place
Catalog No. NC9480421 Supplier Aqua Solutions Supplier No. 86544LC
Sodium Hydroxide 1.5 Normal solution;6%; Clear colorless solution;4L; CAS: 1310-73-2; Store in a cool dry place