Learn More
Hach Company Sodium Hydroxide Digital Titrator Cartridge, 1.600 N
Supplier:  Hach Company 1437901
Encompass
For acidity determination by Digital Titration, 100 - 4000 mg/L CaCO3
Catalog No. 50-199-5010