Learn More
Aqua Solutions Sodium Hydroxide 0.1 Normal NIST Traceable Solution | 1310-73-2 | MFCD00003548 | 20L
SDP
Catalog No. NC1580635
Encompass_Preferred
Sodium Hydroxide 0.1 Normal NIST Traceable Solution | MW: 39.997 | 1310-73-2 | MFCD00003548 | 20L
Catalog No. NC1580635 Supplier Aqua Solutions Supplier No. 862520L
Sodium Hydroxide 0.1 Normal NIST Traceable Solution | MW: 39.997 | 1310-73-2 | MFCD00003548 | 20L