Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sodium Hydroxide, 10.0 N Aqueous Solution, APHA, Spectrum™ Chemical
SDP

Catalog No. 18606548 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-606-548 500 mL
18-606-546 4 L
18-606-547 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-606-548 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S395500ML
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

S-395, 1310-73-2

Spectrum™ Chemical Sodium Hydroxide, 10.0 N Aqueous Solution, APHA - As an APHA certified solution, this item meets the ASTM D1209 standards for use in water and waste water treatment facilities to determine the concentration of dissolved and particulate materials. Spectrum Chemical manufactured APHA grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

CAS 1310-73-2 , 7732-18-5
Molecular Formula HNaO
Molecular Weight (g/mol) 40.00
InChI Key HEMHJVSKTPXQMS-UHFFFAOYSA-M
IUPAC Name sodium hydroxide
SMILES [OH-].[Na+]

Specifications

CAS Max % 0.3, 0.7
Concentration 10 N ± 0.5 N
Quantity 500 mL
Grade APHA
Packaging Plastic
Chemical Name or Material Sodium Hydroxide Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.